Learn More
Cayman Chemical DemethoxyvIrIdIol 5mg
SDP
Supplier:  Cayman Chemical 279535 MG
Encompass_Preferred
A mycotoxin; induces lethality in day-old cockerels (LD50 = 4.2 mg/kg); a PI3K inhibitor
Catalog No. 50-294-1206