Learn More
Medchemexpress LLC Demethylwedelolactone | 6468-55-9 | MFCD09264645 | 99.9% | 300.22 g/mol | C15H8O7 | 10 MG

Supplier: Medchemexpress LLC HYN202810MG
Demethylwedelolactone is a coumestan-class bioactive small molecule supplied for laboratory research as a milligram-scale reagent. It acts as a trypsin inhibitor and is used in biochemical and cell-based studies. The compound has molecular formula C15H8O7, molecular weight 300.22 g/mol, and CAS number 6468-55-9, and is provided as a 10 mg quantity for research use only.
- High reported purity of 99.89%.
- Milligram-scale quantity suitable for assay development and analytical studies.
- Molecular weight 300.22 g/mol and formula C15H8O7 for exact stoichiometry.
- Trypsin inhibitor activity useful in protease-related research.
- CAS number 6468-55-9 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.