Learn More
Medchemexpress LLC HY-N0638 5mg Medchemexpress, Dendrobine CAS:2115-91-5 Purity:>98%
Supplier: Medchemexpress LLC HYN06385MG
Medchemexpress, HY-N0638 5mg Dendrobine CAS:2115-91-5 Dendrobine is an alkaloid isolated from Dendrobium nobile. Dendrobine possesses antiviral activity against influenza A viruses, with IC50s of 3.39 μM, 2.16 μM and 5.32 μM for A/FM-1/1/47 (H1N1), A/Puerto Rico/8/34 H274Y (H1N1) and A/Aichi/2/68 (H3N2), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.