Learn More
Medchemexpress LLC Dendrogenin A | 1191043-85-2 | 96.7% | 513.80 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1491321MG
Encompass_Preferred
Dendrogenin A is a ligand for liver X receptor (LXR) that induces the expression of sodium/iodine symporter, increasing iodine uptake. It also induces cell differentiation of MCF-7 cells, reactivates the function of lactating cells, and influences thyroid hormone generation by inducing expressions of TSH receptor, thyroid peroxidase, and thyroglobulin. Dendrogenin A exhibits cytotoxicity in cancer cells B-CPAP and 8505c, and arrests the cell cycle at the G0/G1 phase.
  • Ligand for liver X receptor (LXR)
  • Induces expression of sodium/iodine symporter
  • Increases iodine uptake
  • Induces cell differentiation of MCF-7 cells
  • Reactivates function of lactating cells
  • Affects thyroid hormone generation
  • Exhibits cytotoxicity in cancer cells
  • Arrests cell cycle at G0/G1 phase
Catalog No. 50-003-59435