Learn More
Medchemexpress LLC Dendrophenol | 108853-14-1 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN60315MG
Encompass_Preferred
Dendrophenol, also known as Moscatilin, is a potent NF-κB inhibitor that exhibits anti-inflammatory properties. It demonstrates significant cytotoxic effects against various tumor cells, leading to cell cycle arrest and apoptosis. This compound also possesses antitumor activity and can inhibit vascular calcification by targeting the WNT3/β-catenin pathway.
  • Inhibits inflammation through NF-κB pathway
  • Exerts cytotoxic effects against tumor cells
  • Induces cell cycle arrest and apoptosis in tumor cells
  • Shows antitumor activity
  • Inhibits vascular calcification by modulating WNT3/β-catenin activation
  • Reduces expression of COX-2, iNOS, and HIF-1α
  • Induces apoptosis in human colon cancer cells
Catalog No. 50-004-18645