Learn More
Medchemexpress LLC Denfivontinib | 1457983-28-6 | MFCD28167815 | 98.0% | 521.41 g/mol | C25H25BrN6O2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY123331MG
Encompass_Preferred
Denfivontinib (G-749) is a small-molecule, ATP-competitive FLT3 inhibitor used in preclinical research. It features a brominated pyrido[4,3-d]pyrimidinone core and has demonstrated antitumor activity in FLT3-driven models. The compound is supplied as a dry powder for laboratory studies; consult safety documents before handling.
Catalog No. 50-002-38106