Learn More
Medchemexpress LLC Denfivontinib | 1457983-28-6 | MFCD28167815 | 98.0% | 521.41 g/mol | C25H25BrN6O2 | 50MG

Supplier: Medchemexpress LLC HY1233350MG
Denfivontinib (G-749) is a potent, orally active ATP-competitive inhibitor of FMS-like tyrosine kinase 3 (FLT3) developed for research applications, including studies of drug resistance in acute myeloid leukemia (AML). It inhibits wild-type and mutant FLT3 at sub-nanomolar concentrations and is supplied as a solid research compound.
- Potent FLT3 inhibition: IC50 0.4 nM (wild type), 0.6 nM (D835Y)
- CAS number: 1457983-28-6
- Chemical formula: C25H25BrN6O2
- Molecular weight: 521.41 g/mol
- Purity: 98.0%
- Physical form: white to off-white solid; hydrochloride salt available
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years)
- Intended use: research tool compound for AML and kinase studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.