Learn More
ATCC Dengue virus 4; Strain: H241; Tissue culture adapted
Supplier:  ATCC VR1490
Encompass
Dengue virus type 4 strain H241 is propagated in rhesus monkey kidney cells (RhMkK) (ATCC CCL-7.1). This product was derived by adaptation of suckling mouse–passaged H241 (ATCC VR-217) to RhMkK tissue culture (ATCC CCL-7.1). This product has applications in vector-borne disease research.
Catalog No. 50-238-2895