Learn More
Medchemexpress LLC Denifanstat | 1399177-37-7 | 99.6% | 439.55 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11282910MM/1ML
Encompass_Preferred
Denifanstat is an orally active and potent Fatty Acid Synthase (FASN) inhibitor. It demonstrates an IC50 of 0.052 μM and an EC50 of 0.072 μM. This compound holds potential for research in fatty liver disease and cancer, as its FASN inhibition mechanism also prevents palmitoylation of SARS-CoV2 spike protein.
  • Orally active and potent Fatty Acid Synthase (FASN) inhibitor.
  • IC50 of 0.052 μM and EC50 of 0.072 μM.
  • Potential for fatty liver disease and cancer research.
  • FASN inhibition prevents palmitoylation of SARS-CoV2 spike protein.
Catalog No. 50-004-37897