Learn More
Medchemexpress LLC Denifanstat | 1399177-37-7 | 99.6% | 439.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112829
Encompass_Preferred
Denifanstat (TVB-2640) is an orally active, potent Fatty Acid Synthase (FASN) inhibitor (IC50 of 0.052 μM, EC50 of 0.072 μM) with potential for fatty liver disease and cancer research. Also known as FASN-IN-2 and ASC-40, it has a molecular weight of 439.55 and formula C27H29N5O. This white to off-white solid is for research use only.
  • Orally active and potent fatty acid synthase (FASN) inhibitor.
  • IC50 of 0.052 μM and EC50 of 0.072 μM for FASN inhibition.
  • Potential for fatty liver disease and cancer research.
  • Prevents palmitoylation of SARS-CoV2 spike protein.
  • Suppresses SARS-CoV-2 virus replication, reduces weight loss, and prolongs survival after SARS-CoV-2 infection.
  • Used in published research for cancer and SARS-CoV-2 studies.
  • Undergoing clinical trials for MASH/NASH, acne vulgaris, and various cancers.
Catalog No. 50-004-39072