Learn More
Medchemexpress LLC Denifanstat (TVB-2640) | 1399177-37-7 | 99.6% | 439.55 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11282910MG
Encompass_Preferred
Denifanstat (TVB-2640) is an orally active and potent Fatty Acid Synthase (FASN) inhibitor with an IC50 of 0.052 μM and an EC50 of 0.072 μM. It has potential for fatty liver disease and cancer research.
  • Orally active and potent FASN inhibitor
  • Potential for fatty liver disease and cancer research
  • FASN inhibition prevents palmitoylation of SARS-CoV2 spike protein.
Catalog No. 50-004-36148