Learn More
Medchemexpress LLC Denopamine | 71771-90-9 | 98.3% | 1 ML

Supplier: Medchemexpress LLC HY11951510MM 1ML
Denopamine is a selective β1-adrenergic agonist supplied as a 10 mM solution in DMSO (1 mL) for research use. It is intended for in vitro and laboratory applications and includes characterization data for purity, molecular weight, and storage recommendations.
- Selective β1-adrenergic agonist suitable for cardiovascular research.
- Supplied as a 10 mM solution in DMSO, 1 mL vial.
- High purity (≈98.3%) with documented analytical data.
- Molecular weight 317.38 g/mol; formula C18H23NO4.
- Soluble in DMSO; warming and sonication improve dissolution.
- Store in solvent at -80°C for long-term stability.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.