Learn More
Anton Paar Density (ASTM D4052, ISO 12185) combined with viscosity in one portable unit. Robust metal cells ensure durability. Only 2 minutes of operator time per test with direct funnel filling—no pipette or syringe needed.
Supplier:  Anton Paar 105013
Encompass_Preferred
Test Methods: ASTM D4052, ISO 12185Measuring Range:Viscosity: 0.3 mm²/s to 5,000 mm²/sDensity: 0 g/cm³ to 3 g/cm³Temperature: +15 °C to +100 °C Density repeatability** and reproducibility**0.00005 g/cm³ and 0.0001 g/cm³Viscosity repeatability** and reproducibility**0.2 % and 0.7 %Temperature repeatability: 0.005 °CTemperature reproducibility: 0.03 °C from +15 °C to +100 °CSample volume min. / typical: 3.5 mL / 8 mLSolvent volume min. / typical: 5 mL / 10 mLMax. sample throughput: 21 samples per hourOptional upgrades: Second temperatureIntegrate Simple Fill Filling Device
Catalog No. 50-005-80440
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.