Learn More
Cayman Chemical Deoxy Donepezil hydrochlorid
SDP
Supplier:  Cayman Chemical 30045500 UG
Encompass_Preferred
A potential impurity found in bulk preparations of donepezil; is produced as a by-product during donepezil synthesis
Catalog No. 50-298-9002