Learn More
Medchemexpress LLC Deoxyarbutin | 53936-56-4 | HY-B1461 | 99.92% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB1461
Encompass_Preferred
Deoxyarbutin is an orally active tyrosinase inhibitor that can promote apoptosis of melanoma cells, enhance the vitality of mouse acinar cells, and has skin whitening and anti-tumor activity.
  • Orally active tyrosinase inhibitor.
  • Promotes apoptosis of melanoma cells.
  • Enhances vitality of mouse acinar cells.
  • Possesses skin whitening activity.
  • Shows anti-tumor activity.
Catalog No. 50-004-14344