Learn More
Selleck Chemical LLC Deoxycholic acid S4689-100mg
SDP
Supplier:  Selleck Chemical LLC S4689100MG
Encompass_Preferred
Deoxycholic acid (Deoxycholate Desoxycholic acid Cholerebic Cholorebic) is a cytolytic agent The physiologic effect of deoxycholic acid is by means of decreased cell membrane integrity Deoxycholic acid is specifically responsible for activating the G protein-coupled bile acid receptor TGR5 that stimulates brown adipose tissue (BAT) thermogenic activity
Catalog No. 50-311-1459