Learn More
Apexbio Technology LLC Deoxycholic acid 83-44-3 20mg
SDP
Supplier:  Apexbio Technology LLC N166920
Encompass_Preferred
Deoxycholic acid (CAS 83-44-3) is a secondary bile acid derived from cholic acid commonly extracted from animal bile sources such as pig bile It acts as a natural surfactant involved in the emulsification and absorption of dietary fats in the intestine by disrupting lipid membranes and facilitating micelle formation Owing to its amphipathic properties deoxycholic acid is widely utilized in studies investigating lipid metabolism cell membrane dynamics and bile acid signaling pathways The compound is soluble in ethanol ether methanol acetone acetic acid and alkaline solutions and is typically stored in a sealed cool and dry environment to maintain stability
Catalog No. 50-000-90656