Learn More
Medchemexpress LLC Deoxycholic acid-d4 | 112076-61-6 | MFCD00209707 | 98.0% | 396.60 | C24H36D4O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0593S10MG
Encompass_Preferred
Deoxycholic acid-d4 is a deuterium-labeled analogue of deoxycholic acid used as an isotope-labeled reference standard in analytical and biochemical applications. It is intended for use in quantitative LC-MS/MS assays, metabolic tracing, and receptor activation studies involving bile acid signaling.
  • Deuterium-labeled (d4) analogue for mass spectrometry internal standard use.
  • High purity suitable for analytical applications, listed at 98.0% purity.
  • Molecular weight approximately 396.60 Da, molecular formula C24H36D4O4.
  • Suitable for quantitative LC-MS/MS and metabolic tracing workflows.
  • Offered in small analytical pack sizes for reference standard use.
Catalog No. 50-002-96407