Learn More
Cayman Chemical DeoxycholIc AcId-d4 5mg
SDP
Supplier:  Cayman Chemical 208515 MG
Encompass_Preferred
An internal standard for the quantification of DCA by GC- or LC-MS
Catalog No. 50-298-3897