Learn More
Medchemexpress LLC HY-N0593A 100mg Medchemexpress, Deoxycholic acid sodium salt CAS:302-95-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0593A100MG
Encompass_Preferred
Medchemexpress, HY-N0593A 100mg Deoxycholic acid sodium salt CAS:302-95-4 Deoxycholic acid sodium salt is specifically responsible for activating the G protein-coupled bile acid receptor TGR5 that stimulates brown adipose tissue (BAT) thermogenic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3628