Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deoxycholic acid (standard) | 83-44-3 | MFCD00003673 | 99.9% | 392.57 | C24H40O4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN0593R10MG

Encompass_Preferred

Deoxycholic acid (Standard) is an analytical reference standard of deoxycholic acid intended for research and analytical applications. Supplied in small packaged quantities, it includes accompanying certificate of analysis and safety data documentation for assay calibration and method development.

  • Analytical standard suitable for assay calibration and method development.
  • Purity 99.85% as specified in the certificate of analysis.
  • CAS number 83-44-3; molecular formula C24H40O4; molecular weight 392.57.
  • Available in small pack sizes (for example, 10 mg) for laboratory use.
  • Accompanied by COA and SDS for quality and safety verification.
  • Store under conditions recommended in the certificate of analysis.

Catalog No. 50-002-96817


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.