Learn More
Cayman Chemical DeoxycortIcosteron Acetate 1g
SDP
Supplier:  Cayman Chemical 308561 G
Encompass_Preferred
A corticosteroid; has commonly been used in rodents as a model of salt-sensitive hypertension, characterized by increases in sympathetic nervous system activity, imbalances in the renin-angiotensin system (RAS), and cardiac remodeling in combination with saline
Catalog No. 50-298-8871