Learn More
Enzo Life Sciences Deoxyfuconojirimycin . hydrochloride (5 mg). CAS: 210174-73-5
SDP
Supplier:  Enzo Life Sciences BMLS1080005
Encompass_Preferred
A potent and selective inhibitor of α-L-fucosidases. Purity: ≥98% (TLC). Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-201-1735