Learn More
Apexbio Technology LLC Deoxyguanosine 5-triphosphate 93919-41-6 10mg
SDP
Supplier:  Apexbio Technology LLC B788310
Encompass_Preferred
Deoxyguanosine 5-triphosphate (CAS 93919-41-6) is a nucleotide analogue that serves as a precursor for DNA synthesis within cells It functions as a substrate for reverse transcriptase facilitating the incorporation of guanine residues during DNA chain elongation Experimental studies demonstrate that its presence enhances DNA amplification efficiency in reverse transcription polymerase chain reaction (RT-PCR) assays Due to these properties Deoxyguanosine 5-triphosphate is widely utilized as a molecular reagent in genomic research and molecular diagnostics supporting investigations into nucleic acid metabolism and gene expression analysis
Catalog No. 50-000-90887