Learn More
Medchemexpress LLC HY-N3733 5mg Medchemexpress, Deoxylapachol CAS:3568-90-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN37335MG
Encompass_Preferred
Medchemexpress, HY-N3733 5mg Deoxylapachol CAS:3568-90-9 Deoxylapachol is a major cytotoxic component of New Zealand brown alga, Landsburgia quercifolia. Deoxylapachol has antifungal and anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2918