Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxynucleoside Triphospha
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DNSTPRO
Encompass
Deoxynucleoside Triphosphate Set consists of four separate vials of dATP dCTP dGTP and dTTP. Each vial contains a 100 mM clear colorless SOLUTIONon of the lithium salt (pH 7).
Catalog No. 50-290-7540