Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxynucleotide Mix 10 mM
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D72955ML
Encompass
Nucleotides are organic molecules that serve as the monomers or subunits of nucleic acids (like DNA and RNA). The building blocks of nucleic acids nucleotides consist of a nitrogenous base (purine or pyrimidine) a five-carbon sugar (ribose or deoxyribose) and at least one phosphate group. A nucleoside along with a phosphate group yields a nucleotide.
Catalog No. 50-291-4723