Learn More
Selleck Chemical LLC Deoxypodophyllotoxin E2017-1MG
SDP
Supplier:  Selleck Chemical LLC E20171MG
Encompass_Preferred
Deoxypodophyllotoxin (DPT) a derivative of podophyllotoxin is a lignin from Anthriscus sylvestris with potent antimitotic anti-inflammatory and antiviral activities
Catalog No. 50-000-40230