Learn More
Cayman Chemical DeoxypodophylotoxIn 10mg
SDP
Supplier:  Cayman Chemical 3643410 MG
Encompass_Preferred
A flavolignan with diverse biological activities; inhibits tubulin polymerization in a cell-free assay at 50 nM; inhibits COX-2 and 5-LO in isolated mouse bone marrow-derived mast cells stimulated with KL, IL-10, and LPS (IC50s = 1.89 and 0.37 µM, respectively); reduces viral yield in the supernatant of HSV-1- or HSV-2-infected MRC-5 cells at 0.02 UG/ml; inhibits aggregation of rabbit platelets induced by PAF, collagen, or arachidonic acid; induces mortality in P. rapae fifth instar larvae (LC50 = 0.02 g/L); reduces tumor growth in a CT26 murine colon cancer model at 70 UG/kg
Catalog No. 50-291-2530