Learn More
Sigma Aldrich Fine Chemicals Biosciences Deoxyribonucleic acid sing
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D91565ML
Encompass
A ready-to-use SOLUTIONon of high quality single-stranded template DNA isolated from the testes of salmon. The SOLUTIONon is supplied at a concentration of 9 - 12 mg/ml .
Catalog No. 50-290-2735