Learn More
Medchemexpress LLC Deoxyribonucleic acid sodium salt from salmon testes | 438545-06-3 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY172445
Encompass_Preferred
Deoxyribonucleic acid sodium salt from salmon testes (ssstDNA) can be used to make anti-adhesion layers (AALs) for passivating gold plasmonic sensor surfaces. It is for research use only.
  • Can be used to make anti-adhesion layers (AALs)
  • Used for passivating gold plasmonic sensor surfaces
  • For research use only
Catalog No. 50-003-85162