Learn More
Medchemexpress LLC Deoxyribonucleic acid sodium salt from salmon testes | 438545-06-3 | MFCD00130922 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17244510MG
Encompass_Preferred
Deoxyribonucleic acid sodium salt from salmon testes is fragmented, double-stranded DNA in the sodium salt form supplied as a dry powder for research use. It is commonly used as a nonspecific blocking agent in hybridization assays and for surface passivation in biochemical and sensor applications. The material is heterogeneous and molecular weight is not precisely defined.
Catalog No. 50-002-95031