Learn More
Medchemexpress LLC Deoxyribonucleic acid sodium salt from salmon testes (SS-DNA) | 438545-06-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1724455MG
Encompass_Preferred
Deoxyribonucleic acid sodium salt from salmon testes (ssstDNA) can be used to make anti-adhesion layers (AALs) for passivating gold plasmonic sensor surfaces. For research use only. Not sold to patients.
Catalog No. 50-003-85392