Learn More
Alkali Scientific DEPC [Diethyl Pyrocarbonate], 100 Milliliters
Supplier: Alkali Scientific DEP001CP
Diethyl Pyrocarbonate (DEPC) is a reagent used to inactivate RNases in laboratory settings, providing an effective means for ensuring the integrity of RNA samples. This 100ml bottle is designed for use in molecular biology applications where RNA must be protected from enzymatic degradation during extraction, handling, or storage. DEPC is widely used in RNA isolation protocols, ensuring that the RNA remains intact for subsequent analyses, including gene expression studies and sequencing. It is commonly used in the preparation of buffers, water, and solutions for RNA experiments to prevent contamination by RNases, which could otherwise interfere with research accuracy.