Learn More
Alkali Scientific DEPC [Diethyl Pyrocarbonate], 200 Milliliters
SDP
Supplier:  Alkali Scientific DEP002CP
Encompass
Diethyl Pyrocarbonate (DEPC) is a powerful RNase inhibitor used in molecular biology to protect RNA from degradation. Provided in a 200ml bottle, DEPC is ideal for use in laboratories working with RNA extraction, cDNA synthesis, and other RNA-related experiments. DEPC works by inactivating RNases, enzymes that could otherwise degrade RNA during handling and analysis. It is commonly used to treat water, buffers, and solutions prior to RNA isolation to ensure the integrity of the RNA sample. This reagent plays a crucial role in maintaining RNA sample purity, ensuring accurate and reliable results in gene expression studies, RNA sequencing, and other molecular research.
Catalog No. 50-006-61121