Learn More
Medchemexpress LLC Depressine | 176182-06-2 | 99.8% | 688.63 g·mol-1 | C30H40O18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN507010MG
Encompass_Preferred
Depressine is an iridoid glycoside isolated from the aerial parts of Gentiana depressa and used as a DNA protectant to reduce oxidative DNA base damage and strand breaks, for example during silver nanoparticle preparation.
  • Iridoid glycoside isolated from Gentiana depressa.
  • Reduces oxidative DNA base damage and strand breaks.
  • Useful for protecting DNA during silver nanoparticle preparation.
  • High purity (99.8%).
  • Molecular weight 688.63 g·mol-1; chemical formula C30H40O18.
Catalog No. 50-002-89704