Learn More
Medchemexpress LLC Deprodone propionate | 20424-00-4 | 400.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00190100MG
Encompass_Preferred
Deprodone propionate is a corticosteroid obtained by esterifying the 17-position of the prednisolone skeleton with propionic acid and deoxidating its 21-position. It is intended for research use only. Not for sale to patients. Deprodone propionate can exhibit its effects to the maximum extent and over a prolonged period of time, does not cause skin irritation, and is physicochemically stable. It can be maintained in a stably suspended state due to the thickening action of the suspending agent.
  • Effective ingredient with prolonged action
  • Does not cause skin irritation
  • Physicochemically stable
  • Maintained in a stably suspended state
Catalog No. 50-004-47632