Learn More
Medchemexpress LLC Deprodone propionate (RD20000) | 20424-00-4 | 400.51 | 50 MG

Supplier: Medchemexpress LLC HYU0019050MG
Deprodone propionate (RD20000) is a corticosteroid obtained by esterifying the 17-position of the prednisolone skeleton with propionic acid and deoxidating its 21-position. It is intended for research use only.
- Corticosteroid compound
- Obtained by esterifying the 17-position of the prednisolone skeleton with propionic acid
- Deoxidating the 21-position
- Appears as a solid, white to off-white in color
- Purity of 99.62%
- Soluble in DMSO at 250 mg/mL
- Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
- Recommended storage in solvent: -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.