Learn More
MSC Mitutoyo Depth Probe, For Linear Height Gages, 94.6 mm Vertical Length, 14 mm Shank, 1 ea
Supplier: MSC 08967911
This depth probe is designed to extend the utility of linear height gages, enabling precise measurement of complicated workpiece profiles and various measurement functions. It is compatible with Mitutoyo LH-600 and QM-Height series linear height gages.
- Compatible with linear height gages including LH-600 and QM-Height
- Enables measurement of complex workpiece profiles
- Features a 94.6 mm vertical length and 14 mm shank size
- Supports attachment of various coordinate measuring machine (CMM) styli
- Equipped with an M2/M3 threaded shank for versatile probe integration