Learn More
Cytiva (Formerly Pall Lab) Supracap™ 100 depth filter capsule, in-line style, 5 inch length, with single-layer media grade K200P (3-6 m retention rating), 1 to 1 inch sanitary flange inlet and outlet
Supplier:  Cytiva NP5LP2001
Encompass
Scalable Capsule with Seitz High Performance Depth Filter Media-Flexible choices in filter media for prefiltration and removal of coarse particulate.-Low hold-up volume allows for increased product recovery and requires low post-use rinse volumes.-For filtration of 3 to 100 L.-Mechanically robust design provides unobstructed process flows, consistent and scalable filtration results, and high filter media integrity. -Completely disposable technology -Encapsulated design reduces operator exposure to potential biohazards. -Tested and certified. Manufactured under a Quality Management System certified to ISO 9000 and ISO 9001, and an Environmental Management System certified to ISO 14001.-Offers flexibility and assurance of application success from development to production scale. Scalable to Palls entire line of traditional SUPRAdisc™ modules, as well as Stax™ capsules. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1613238