Learn More
TARGETMOL CHEMICALS INC DEPTOR-IN-1 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T402421MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.DEPTOR-IN-1 is an inhibitor of DEPTOR with a Kd of 9.3 uM. Purity 97.84%
Catalog No. 50-287-5807