Learn More
Selleck Chemical LLC Dequalinium Chloride S4066-50mg
SDP
Supplier:  Selleck Chemical LLC S406650MG
Encompass_Preferred
Dequalinium Chloride is a PKC inhibitor with IC50 of 7-18 M and also a selective blocker of apamin-sensitive K channels with IC50 of 1 1 M
Catalog No. 50-310-9703