Learn More
Cayman Chemical DequalInIumchlorIde 250mg
SDP
Supplier:  Cayman Chemical 17557250 MG
Encompass_Preferred
An ammonium-containing cationic surfactant with diverse biological activities; has antifungal, antitrichomonal, and antibacterial activity; inhibits apamin binding to and net potassium loss mediated by SKCa channels in angiotensin II-stimulated guinea pig hepatocytes; inhibits primary and recurrent tumor growth in a W163 rat colon carcinoma isograft model from 1-10 mg/kg
Catalog No. 50-296-7251