Learn More
Medchemexpress LLC Der p 23 protein (Dermatophagoides pteronyssinus) | >95.0% | 5 UG

Supplier: Medchemexpress LLC HYP752895UG
Recombinant Der p 23 is the mature major allergen from the house dust mite Dermatophagoides pteronyssinus, provided lyophilized for laboratory use. The preparation corresponds to amino acids A22-T90, is expressed in HEK293 cells with a C-terminal His tag, and is supplied at 5 μg with >95% purity. It is intended for immunoassays, antibody production, and allergen characterization.
- Expressed in HEK293 cells for eukaryotic folding and modifications.
- C-terminal His tag facilitates detection and purification.
- Purity >95.0% as determined by reducing SDS-PAGE.
- Contains the mature region A22-T90, approx. 19 kDa.
- Lyophilized from PBS, pH 7.4, for increased stability.
- Supplied as 5 μg aliquots suitable for analytical assays.
- Store lyophilized at -20°C; aliquot and freeze for long-term storage.
- Suitable for immunoassays and antibody generation applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.