Learn More
Key Scientific Products DERMAPAK 2000 - BLACK, 100 per Pack
SDP
Supplier:  Key Scientific Products DMK201
Encompass_Preferred
Compact, lightweight: low mailing costs. Safe for transporting specimen. Absorbent action reduce risk of bacterial overgrowth during transport. Used by Clinician and Lab. Established, proven design. Secure, robust, customizable check panel.
Catalog No. NC9491419