Learn More
Medchemexpress LLC Dermaseptin | 136212-91-4 | 99.7% | 3455.10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP02635MG
Encompass_Preferred
Dermaseptin is a peptide isolated from frog skin that exhibits potent antimicrobial activity against bacteria, fungi, and protozoa at micromolar concentration. It is a water-soluble, thermostable, and nonhemolytic peptide with highly potent antimicrobial activity against pathogenic fungi at micromolar concentrations.
  • Exerts lytic action on bacteria, protozoa, yeasts, and filamentous fungi.
  • Mechanism involves an amphipathic alpha-helix perturbing membrane functions.
  • Broad spectrum: Inhibits 100% proliferation of most microorganisms (5-100 μg/ml).
  • Low in vitro toxicity to human cells, not inhibiting KJ3 cell proliferation or permeating guinea pig lymphocytes.
  • Hemolysis of rabbit erythrocytes observed at doses above 200 μg/ml.
  • Active against *Aeromonas cauiae*, *Pseudomonas aeroginusa*, *Escherichia coli*, *Enterococcus faecalis*, *L. mezicana*, and *Microsporum canis*.
Catalog No. 50-004-25357