Learn More
Medchemexpress LLC Dermaseptin TFA | 646451-06-1 | 99.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP0263A5MG
Encompass_Preferred
Dermaseptin TFA is a peptide isolated from frog skin, known for its potent antimicrobial activity against various microorganisms. It is a water-soluble, thermostable, and nonhemolytic peptide.
  • Exhibits potent antimicrobial activity against bacteria, fungi, and protozoa
  • Inhibits proliferation of most tested microorganisms at micromolar concentrations
  • Functions through an NH2-terminal alpha-helical amphipathic segment
  • Stable under recommended storage conditions
Catalog No. 50-004-26011