Learn More
Selleck Chemical LLC Deruxtecan S9795-5MG
SDP
Supplier:  Selleck Chemical LLC S97955MG
Encompass_Preferred
Deruxtecan is an ADC drug-linker conjugate composed of the cleavable glycine-glycine-phenylalanine-glycine tetrapeptide-based linker a self-immolative amino methylene spacer and a novel topoisomerase 1 inhibitor payload that is a derivative of exatecan (DX-8951)
Catalog No. 50-000-23832