Learn More
Medchemexpress LLC Deruxtecan-d5 | 1599440-13-7 | 98.4% | 1039.08 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13631ES450MG
Encompass_Preferred
Deruxtecan-d5 is a deuterium-labeled compound, an ADC drug-linker conjugate that integrates a DX-8951 derivative (DXd) and a maleimide-GGFG peptide linker. This compound is instrumental in the synthesis of DS-8201 and U3-1402.
  • Can be used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-51783