Learn More
MOLECULAR DEPOT LLC Des-Arg9-Bradykinin - 5 mg
Supplier:  MOLECULAR DEPOT LLC MDP0427
Encompass_Preferred
Des-Arg9-Bradykinin Peptide is a synthetic peptide consisting of the sequence Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe The molecular weight of this peptide is 904 02 g/mol and the CAS number is 15958-92-6 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13392